Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO1C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145944.1 | NP_001139416.1 | 5, 6 | 851 | ||
NM_001145945.1 | NP_001139417.1 | 7, 8 | 987 | |||
NM_001145946.1 | NP_001139418.1 | 8, 9 | 1134 | |||
NM_017435.4 | NP_059131.1 | 7, 8 | 1147 | |||
XM_005253394.2 | XP_005253451.1 | 8, 9 | 923 | |||
XM_005253396.2 | XP_005253453.1 | 6, 7 | 5688 | |||
XM_011520703.2 | XP_011519005.1 | 8, 9 | 923 | |||
XM_011520704.2 | XP_011519006.1 | 8, 9 | 923 | |||
XM_011520709.2 | XP_011519011.1 | 5, 6 | 565 | |||
XM_011520711.2 | XP_011519013.1 | 7, 8 | 5743 | |||
XM_017019484.1 | XP_016874973.1 | 10, 11 | 6184 | |||
XM_017019485.1 | XP_016874974.1 | 9, 10 | 6102 | |||
XM_017019486.1 | XP_016874975.1 | 7, 8 | 5842 | |||
XM_017019487.1 | XP_016874976.1 | 6, 7 | 5688 | |||
XM_017019488.1 | XP_016874977.1 | 11, 12 | 6239 | |||
XM_017019489.1 | XP_016874978.1 | 12, 13 | 6339 | |||
XM_017019490.1 | XP_016874979.1 | 12, 13 | 6303 | |||
GenBank mRNA | AF205076.1 | - | 7, 8 | 1148 | ||
AF260704.1 | - | 7, 8 | 843 | |||
AK294333.1 | - | 5, 6 | 719 | |||
AK296236.1 | - | 7, 8 | 880 | |||
AK303713.1 | - | 8, 9 | 1010 | |||
AK308729.1 | - | 6, 7 | 783 | |||
AL834209.1 | - | 8, 9 | 999 | |||
BC022461.2 | - | 7, 8 | 876 | 4801171 |