Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PPIL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032472.3 | NP_115861.1 | 5, 6 | 501 | ||
NM_130906.2 | NP_570981.1 | 5, 6 | 489 | |||
XM_005246651.3 | XP_005246708.1 | 5, 6 | 420 | |||
XM_005246652.4 | XP_005246709.1 | 5, 6 | 408 | |||
XM_011511358.2 | XP_011509660.1 | 5, 6 | 594 | |||
XM_017004352.1 | XP_016859841.1 | 5, 6 | 611 | |||
XM_017004353.1 | XP_016859842.1 | 5, 6 | 350 | |||
XM_017004354.1 | XP_016859843.1 | 5, 6 | 784 | |||
XM_017004355.1 | XP_016859844.1 | 5, 6 | 808 | |||
XM_017004356.1 | XP_016859845.1 | 5, 6 | 772 | |||
XM_017004357.1 | XP_016859846.1 | 6, 7 | 691 | |||
XM_017004358.1 | XP_016859847.1 | 6, 7 | 514 | |||
XM_017004359.1 | XP_016859848.1 | 6, 7 | 698 | |||
XM_017004360.1 | XP_016859849.1 | 4, 5 | 511 | |||
GenBank mRNA | AF146799.1 | - | 5, 6 | 358 | ||
AF251049.1 | - | 5, 6 | 501 | |||
AF271652.1 | - | 5, 6 | 424 | |||
AF414127.1 | - | 4, 5 | 425 | |||
AK297804.1 | - | 5, 6 | 420 | |||
AY257468.1 | - | 4, 5 | 545 | |||
BC007693.2 | - | 5, 6 | 930 | 3956681 | ||
CB961908.1 | - | 5, 6 | 544 | |||
KU178646.1 | - | 3, 4 | 248 | |||
KU178647.1 | - | 4, 5 | 301 | |||
KU178648.1 | - | 3, 4 | 260 |