Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: GPR88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022049.2 | NP_071332.2 | 2 | 590 | ||
GenBank mRNA | AB042410.1 | - | 2 | 623 | ||
AK225748.1 | - | 1 | 252 | |||
BC036775.1 | - | 2 | 590 |
Set Membership: |
![]() |