Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOX18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018419.2 | NP_060889.1 | 2 | 1617 | ||
GenBank mRNA | AF086136.1 | - | 453 | |||
AF270652.1 | - | 2 | 1629 | |||
AJ243896.1 | - | 2 | 1519 | |||
BC035513.1 | - | 2 | 1617 | |||
BC038590.1 | - | 2 | 1619 | 5589289 | ||
BC073971.1 | - | 2 | 1619 | 6183010 | ||
BC111390.1 | - | 2 | 1617 | 5751646 |