Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018982.4 | NP_061855.1 | 7 | 821 | ||
NR_036639.1 | 7 | 821 | ||||
NR_036640.1 | 6 | 601 | ||||
NR_135075.1 | 5 | 538 | ||||
XM_017001499.1 | XP_016856988.1 | 6 | 579 | |||
XM_017001500.1 | XP_016856989.1 | 6 | 642 | |||
XM_017001501.1 | XP_016856990.1 | 5 | 423 | |||
GenBank mRNA | AK300240.1 | - | 6 | 683 | ||
AK315183.1 | - | 7 | 799 | |||
AL713668.1 | - | 6 | 532 | |||
BC009674.1 | - | 2 | 134 | |||
BC064590.1 | - | 7 | 787 | 5749036 | ||
BI757212.1 | - | 7 | 816 | |||
BX405963.2 | - | 7 | 813 |