Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018983.3 | NP_061856.1 | 4 | 694 | ||
NM_032993.2 | NP_127460.1 | 4 | 435 | |||
GenBank mRNA | AJ276003.1 | - | 4 | 651 | ||
AK310489.1 | - | 4 | 577 | |||
AY780787.1 | - | 3 | 387 | |||
BC003413.1 | - | 4 | 435 | 3451136 |