Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SASH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018990.3 | NP_061863.1 | 2 | 323 | ||
XM_006724763.1 | XP_006724826.1 | 2 | 290 | |||
GenBank mRNA | AK292023.1 | - | 2 | 271 | ||
AK296665.1 | - | 2 | 225 | |||
AK314869.1 | - | 2 | 290 | |||
AL049683.1 | - | 2 | 248 | |||
BC051881.1 | - | 2 | 255 | 6011968 | ||
CR975883.1 | - | 2 | 323 |