Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APBB1IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019043.3 | NP_061916.3 | 13 | 1588 | ||
XM_006717451.2 | XP_006717514.1 | 12 | 1286 | |||
XM_011519514.2 | XP_011517816.1 | 12 | 1457 | |||
GenBank mRNA | AK303597.1 | - | 13 | 2148 | ||
AY152730.1 | - | 12 | 1354 | |||
BC054516.1 | - | 12 | 1366 | 5172946 |