Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASNSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019048.2 | NP_061921.1 | 5 | 2020 | ||
XM_017004381.1 | XP_016859870.1 | 4 | 1751 | |||
XM_017004382.1 | XP_016859871.1 | 4 | 2536 | |||
GenBank mRNA | AB075481.1 | - | 2 | 1469 | ||
AK000759.1 | - | 5 | 1999 | |||
AY116969.1 | - | 2 | 1586 | |||
BC001243.1 | - | 5 | 1925 | 3453136 |