Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195215.1 | NP_001182144.1 | 20 | 1833 | ||
NR_125340.1 | 17 | 1580 | ||||
XM_005244931.2 | XP_005244988.1 | 19 | 1773 | |||
XM_006711192.2 | XP_006711255.1 | 21 | 1743 | |||
XM_006711193.3 | XP_006711256.1 | 21 | 1875 | |||
XM_006711194.3 | XP_006711257.1 | 8 | 11871 | |||
XM_011509246.2 | XP_011507548.1 | 20 | 1694 | |||
XM_011509247.1 | XP_011507549.1 | 19 | 1634 | |||
XM_011509248.2 | XP_011507550.1 | 20 | 1627 | |||
XM_011509249.2 | XP_011507551.1 | 21 | 1932 | |||
XM_017000470.1 | XP_016855959.1 | 21 | 1821 | |||
XM_017000471.1 | XP_016855960.1 | 20 | 1683 | |||
XM_017000472.1 | XP_016855961.1 | 16 | 1307 | |||
GenBank mRNA | AK000061.1 | - | 6 | 370 | ||
AK091207.1 | - | 17 | 1394 | |||
BC074735.2 | - | 3 | 219 | |||
BP220095.1 | - | 6 | 370 |