Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019050.2 | NP_061923.2 | 10 | 1850 | ||
XM_005263073.3 | XP_005263130.1 | 9 | 1724 | |||
XM_005263075.2 | XP_005263132.1 | 10 | 1889 | |||
XM_005263076.2 | XP_005263133.1 | 10 | 1889 | |||
XM_005263077.3 | XP_005263134.1 | 10 | 1889 | |||
XM_006714240.1 | XP_006714303.1 | 8 | 925 | |||
XM_011532037.2 | XP_011530339.1 | 10 | 1902 | |||
XM_011532038.2 | XP_011530340.1 | 9 | 1179 | |||
XM_017008312.1 | XP_016863801.1 | 11 | 2016 | |||
XM_017008313.1 | XP_016863802.1 | 10 | 1701 | |||
XM_017008314.1 | XP_016863803.1 | 10 | 1700 | |||
XM_017008315.1 | XP_016863804.1 | 10 | 1770 | |||
XM_017008316.1 | XP_016863805.1 | 9 | 1741 | |||
XM_017008317.1 | XP_016863806.1 | 9 | 1713 | |||
XM_017008318.1 | XP_016863807.1 | 10 | 1835 | |||
GenBank mRNA | AB037771.1 | - | 3 | 299 | ||
AJ583824.1 | - | 7 | 781 | |||
BX648339.1 | - | 7 | 904 | |||
CR749490.1 | - | 11 | 1964 |