Gene Symbol: DDX49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019070.4 | NP_061943.2 | 8 | 965 | ||
NR_033677.1 | 8 | 894 | ||||
XM_011528083.1 | XP_011526385.1 | 9 | 1013 | |||
XM_011528084.2 | XP_011526386.1 | 9 | 957 | |||
GenBank mRNA | AK001294.1 | - | 8 | 913 | ||
AK223294.1 | - | 8 | 908 | |||
AK291916.1 | - | 8 | 925 | |||
AL110272.1 | - | 4 | 1031 | |||
AL137702.1 | - | 4 | 1031 | |||
BC000979.3 | - | 9 | 989 | 2901163 | ||
BC002674.2 | - | 8 | 955 | 3607922 | ||
BX647408.1 | - | 9 | 832 |