Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CENPF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016343.3 | NP_057427.3 | 19 | 9177 | ||
XM_011509082.2 | XP_011507384.1 | 18 | 9008 | |||
XM_017000086.1 | XP_016855575.1 | 19 | 9164 | |||
GenBank mRNA | BG680819.1 | - | 4 | 711 | ||
U19769.1 | - | 9461 | ||||
U25725.1 | - | 7 | 2714 | |||
U30872.1 | - | 19 | 9072 |