Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT1A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019078.1 | NP_061951.1 | 1 | 193 | ||
GenBank mRNA | AY435140.1 | - | 1 | 193 |