Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CETN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004066.2 | NP_004057.1 | 1 | 505 | ||
GenBank mRNA | AK312047.1 | - | 1 | 468 | ||
BC029515.1 | - | 1 | 491 | 5297994 | ||
U03270.1 | - | 1 | 481 |