Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099658.1 | NP_001093128.1 | 3 | 1666 | ||
NM_001099660.1 | NP_001093130.1 | 4 | 1773 | |||
NM_018334.4 | NP_060804.3 | 2 | 1584 | |||
GenBank mRNA | AB060967.1 | - | 1 | 1033 | ||
AF134481.1 | - | 1 | 1064 | |||
AK001991.1 | - | 4 | 1685 | |||
AK292556.1 | - | 2 | 1173 | |||
AL442092.1 | - | 1 | 989 | |||
AY358280.1 | - | 3 | 1206 | |||
BC035133.2 | - | 2 | 1463 | 5263900 | ||
CR457397.1 | - | 1 | 727 |