Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM20A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243746.1 | NP_001230675.1 | 11, 12 | 1872 | ||
NM_017565.3 | NP_060035.2 | 10, 11 | 1646 | |||
NR_027751.1 | 10, 11 | 1073 | ||||
XM_006721959.2 | XP_006722022.1 | 10, 11 | 1018 | |||
GenBank mRNA | AB545606.1 | - | 10, 11 | 1646 | ||
AF447872.1 | - | 4, 5 | 1459 | |||
AK056789.1 | - | 11, 12 | 1486 | |||
AK298071.1 | - | 11, 12 | 1514 | |||
AL133105.1 | - | 8, 9 | 755 | |||
AY358197.1 | - | 11, 12 | 1646 | |||
BC036222.1 | - | 10, 11 | 1037 | |||
BC136686.1 | - | 10, 11 | 1535 | 9020676 | ||
BC136689.1 | - | 11, 12 | 1535 | 9020679 |