Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZRANB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017580.2 | NP_060050.2 | 1 | 1063 | ||
XM_005269925.4 | XP_005269982.1 | 2 | 918 | |||
XM_005269926.4 | XP_005269983.1 | 2 | 1106 | |||
XM_006717907.2 | XP_006717970.1 | 2 | 1788 | |||
XM_017016357.1 | XP_016871846.1 | 2 | 2519 | |||
XM_017016358.1 | XP_016871847.1 | 1 | 631 | |||
GenBank mRNA | AJ252060.1 | - | 1 | 1098 | ||
AK302810.1 | - | 2 | 874 | |||
AK307386.1 | - | 1 | 703 | |||
AL832925.1 | - | 1 | 243 |