Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TRPM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195227.1 | NP_001182156.1 | 21 | 3049 | ||
NM_001321281.1 | NP_001308210.1 | 20 | 3157 | |||
NM_001321282.1 | NP_001308211.1 | 21 | 3266 | |||
NM_001321283.1 | NP_001308212.1 | 20 | 3146 | |||
NM_001321285.1 | NP_001308214.1 | 17 | 2769 | |||
NM_017636.3 | NP_060106.2 | 22 | 3484 | |||
XM_005259017.1 | XP_005259074.1 | 16 | 2577 | |||
GenBank mRNA | AF497623.1 | - | 22 | 3448 | ||
AJ575813.1 | - | 22 | 3417 | |||
AK000048.1 | - | 20 | 3177 | |||
AK000235.1 | - | 11 | 2340 | |||
AK292862.1 | - | 22 | 3437 | |||
AK295820.1 | - | 17 | 2668 | |||
AK307897.1 | - | 20 | 3159 | |||
AY046396.1 | - | 20 | 3076 | |||
AY297044.1 | - | 20 | 3072 | |||
AY297045.1 | - | 22 | 3428 | |||
AY297046.1 | - | 21 | 2993 | |||
BC132727.1 | - | 22 | 3411 | 40146749 | ||
BC144123.1 | - | 20 | 3473 |