Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BNC2 SIRNA Assays ›
Gene Symbol:
BNC2
Gene Name:
basonuclin 2
Gene Aliases:
BSN2
Chromosome Location:
Chr.9: 16409503 - 16870838 on Build GRCh38
UniGene ID:
Hs.656581
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s29451
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BNC2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001317939.1
NP_001304868.1
5
945
NM_001317940.1
NP_001304869.1
5
851
NM_017637.5
NP_060107.3
6
1071
XM_011517924.1
XP_011516226.1
5
995
XM_011517934.2
XP_011516236.1
3
2023
XM_017014816.1
XP_016870305.1
8
3381
XM_017014817.1
XP_016870306.1
7
1530
XM_017014818.1
XP_016870307.1
6
1445
XM_017014819.1
XP_016870308.1
7
1529
XM_017014820.1
XP_016870309.1
7
1529
XM_017014821.1
XP_016870310.1
6
1443
XM_017014822.1
XP_016870311.1
4
991
XM_017014823.1
XP_016870312.1
4
5760
XM_017014824.1
XP_016870313.1
3
1312
XM_017014825.1
XP_016870314.1
7
1528
XM_017014826.1
XP_016870315.1
7
1528
XM_017014827.1
XP_016870316.1
7
1528
XM_017014828.1
XP_016870317.1
7
1527
XM_017014829.1
XP_016870318.1
5
1499
GenBank mRNA
AK131398.1
-
6
953
AK299073.1
-
5
821
AK304745.1
-
3
613
AY438376.1
-
4
716
BX641030.1
-
1
102
CR933649.1
-
5
904
DQ884938.1
-
7
1061
DQ884939.1
-
7
1057
DQ884940.1
-
6
1045
DQ884941.1
-
7
1034
DQ884944.1
-
4
970
Back To Top
More Information
Panther Classification:
Molecular Function -
zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
endochondral bone growth
transcription, DNA-templated
regulation of transcription, DNA-templated
tongue development
palate development
mesenchyme development
DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-67fd5c5bdd-kprhh:80/10.239.160.57:80.
git-commit: 7fb76fe7d0d673187a4c374cd16a9abd242c4866
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.3-offline