Search Thermo Fisher Scientific
Gene Symbol: CEACAM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291485.1 | NP_001278414.1 | 3 | 575 | ||
NM_006890.4 | NP_008821.2 | 3 | 575 | |||
GenBank mRNA | AF006622.1 | - | 3 | 466 | ||
AK292213.1 | - | 3 | 568 | |||
AK296634.1 | - | 4 | 505 | |||
AK311285.1 | - | 3 | 403 | |||
AK313108.1 | - | 3 | 568 | |||
BC121132.2 | - | 3 | 469 | 40120888 | ||
L31792.1 | - | 3 | 465 | |||
X98311.1 | - | 3 | 465 |