Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267.2 | NP_001258.2 | 2 | 867 | ||
XM_011524214.2 | XP_011522516.1 | 2 | 1882 | |||
GenBank mRNA | AF371328.1 | - | 2 | 880 | ||
AK292177.1 | - | 2 | 910 | |||
BC036360.1 | - | 2 | 865 | 5163623 | ||
BC073974.1 | - | 2 | 867 | 5730623 |