Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PIGG SIRNA Assays ›
Gene Symbol:
PIGG
Gene Name:
phosphatidylinositol glycan anchor biosynthesis class G
Gene Aliases:
GPI7, LAS21, MRT53, PRO4405, RLGS1930
Chromosome Location:
Chr.4: 499200 - 539921 on Build GRCh38
UniGene ID:
Hs.7099
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s29638
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PIGG
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001127178.2
NP_001120650.1
7
1296
NM_001289051.1
NP_001275980.1
7
1105
NM_001289052.1
NP_001275981.1
5
897
NM_001289053.1
NP_001275982.1
7
1854
NM_001289055.1
NP_001275984.1
8
1366
NM_017733.4
NP_060203.3
7
1296
NR_110293.1
7
1296
XM_005272284.2
XP_005272341.1
7
1530
XM_011513490.2
XP_011511792.1
7
1259
XM_011513491.1
XP_011511793.1
8
1326
XM_011513492.1
XP_011511794.1
2
180
XM_011513493.1
XP_011511795.1
7
1296
XM_011513494.2
XP_011511796.1
7
1296
XM_017008321.1
XP_016863810.1
7
1530
XM_017008322.1
XP_016863811.1
6
1388
XM_017008323.1
XP_016863812.1
8
1668
XM_017008324.1
XP_016863813.1
7
1530
XM_017008325.1
XP_016863814.1
8
1381
XM_017008326.1
XP_016863815.1
7
1296
GenBank mRNA
AB162713.1
-
7
1266
AK000272.1
-
2
62
AK027465.1
-
1
15
AK074715.1
-
7
1068
AK074815.1
-
7
1275
AK296507.1
-
8
1357
AY358538.1
-
7
1238
BC001249.1
-
3
298
BC050262.1
-
7
1766
BC110878.1
-
5
839
5750347
BC144576.1
-
7
1253
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
GPI anchor biosynthetic process
preassembly of GPI anchor in ER membrane
phosphotransferase activity, for other substituted phosphate groups
CP2 mannose-ethanolamine phosphotransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-xbcvc:80/10.239.164.135:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline