Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RCBTB2 SIRNA Assays ›
Gene Symbol:
RCBTB2
Gene Name:
RCC1 and BTB domain containing protein 2
Gene Aliases:
CHC1L, RLG
Chromosome Location:
Chr.13: 48488963 - 48535995 on Build GRCh38
UniGene ID:
Hs.652712
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s2966
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RCBTB2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001268.3
NP_001259.1
5
627
NM_001286830.1
NP_001273759.1
4
562
NM_001286831.1
NP_001273760.1
2
367
NM_001286832.1
NP_001273761.1
2
367
XM_005266242.2
XP_005266299.1
4
480
XM_005266244.2
XP_005266301.1
3
415
XM_006719759.3
XP_006719822.1
4
448
XM_011534894.2
XP_011533196.1
3
375
XM_011534895.1
XP_011533197.1
2
239
XM_017020369.1
XP_016875858.1
6
901
XM_017020370.1
XP_016875859.1
5
773
XM_017020371.1
XP_016875860.1
4
646
XM_017020372.1
XP_016875861.1
3
256
XM_017020373.1
XP_016875862.1
2
438
XM_017020374.1
XP_016875863.1
6
901
XM_017020375.1
XP_016875864.1
6
901
XM_017020376.1
XP_016875865.1
6
901
XM_017020377.1
XP_016875866.1
6
901
XM_017020378.1
XP_016875867.1
5
576
XM_017020379.1
XP_016875868.1
4
511
GenBank mRNA
AB209543.1
-
2
333
AF060219.1
-
4
408
AK023010.1
-
4
370
AK122778.1
-
2
438
AK298438.1
-
3
287
AK301516.1
-
4
386
AK304598.1
-
2
191
AK310081.1
-
2
197
AK315703.1
-
5
547
BC029052.1
-
5
431
5194795
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of GTPase activity
Ran guanyl-nucleotide exchange factor activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0