Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHCHD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317177.1 | NP_001304106.1 | 7 | 714 | ||
NM_017812.3 | NP_060282.1 | 6 | 699 | |||
NR_133671.1 | 7 | 789 | ||||
GenBank mRNA | AK000427.1 | - | 6 | 615 | ||
AK129533.1 | - | 6 | 580 | |||
AK310236.1 | - | 7 | 701 | |||
AL516679.3 | - | 7 | 611 | |||
BC011596.2 | - | 6 | 621 | 3346346 | ||
BC014839.1 | - | 6 | 623 | 2901125 |