Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAUS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166269.1 | NP_001159741.1 | 8 | 1072 | ||
NM_001166270.1 | NP_001159742.1 | 7 | 929 | |||
NM_017815.2 | NP_060285.2 | 8 | 1064 | |||
GenBank mRNA | AK000431.1 | - | 8 | 1033 | ||
AK026016.1 | - | 8 | 1018 | |||
AK057779.1 | - | 6 | 859 | |||
AY927599.1 | - | 3 | 312 | |||
BC001916.1 | - | 8 | 1061 | 2905474 | ||
BC002554.2 | - | 8 | 995 | 3139198 | ||
BX247981.1 | - | 5 | 870 | |||
BX247995.1 | - | 7 | 881 | |||
BX248256.1 | - | 7 | 896 |