Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPCAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017839.4 | NP_060309.2 | 14 | 1691 | ||
XM_011523169.2 | XP_011521471.1 | 14 | 1591 | |||
XM_017023376.1 | XP_016878865.1 | 15 | 1665 | |||
GenBank mRNA | AB244718.1 | - | 14 | 1510 | ||
AK000488.1 | - | 14 | 1618 | |||
AK308386.1 | - | 14 | 1619 | |||
BC002472.2 | - | 14 | 1650 | 3347690 | ||
BX641069.1 | - | 9 | 1593 |