Gene Symbol: TRNAU1AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017846.4 | NP_060316.1 | 5 | 436 | ||
NR_003109.1 | 6 | 584 | ||||
XM_017001542.1 | XP_016857031.1 | 4 | 610 | |||
GenBank mRNA | AK000510.1 | - | 5 | 368 | ||
AK023555.1 | - | 5 | 1016 | |||
AK300162.1 | - | 6 | 760 | |||
AK309481.1 | - | 6 | 516 | |||
BC000680.2 | - | 5 | 360 | 3349210 | ||
BC035855.1 | - | 4 | 960 | |||
BC039879.1 | - | 6 | 502 | 5769935 | ||
BI602122.1 | - | 5 | 441 |