Gene Symbol: TRNAU1AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017846.4 | NP_060316.1 | 7 | 652 | ||
NR_003109.1 | 8 | 800 | ||||
XM_017001542.1 | XP_016857031.1 | 6 | 826 | |||
GenBank mRNA | AK000510.1 | - | 7 | 584 | ||
BC000680.2 | - | 7 | 576 | 3349210 | ||
BC035855.1 | - | 6 | 1176 | |||
BC039879.1 | - | 8 | 718 | 5769935 | ||
BI602122.1 | - | 7 | 657 |