Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf109 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303030.1 | NP_001289959.1 | 3, 4 | 836 | ||
NM_001303031.1 | NP_001289960.1 | 3, 4 | 839 | |||
NM_017850.2 | NP_060320.1 | 3, 4 | 718 | |||
XM_017001546.1 | XP_016857035.1 | 3, 4 | 1904 | |||
XM_017001547.1 | XP_016857036.1 | 3, 4 | 987 | |||
XM_017001548.1 | XP_016857037.1 | 5, 6 | 1109 | |||
XM_017001549.1 | XP_016857038.1 | 4, 5 | 986 | |||
XM_017001550.1 | XP_016857039.1 | 4, 5 | 989 | |||
XM_017001551.1 | XP_016857040.1 | 3, 4 | 655 | |||
XM_017001552.1 | XP_016857041.1 | 5, 6 | 991 | |||
XM_017001553.1 | XP_016857042.1 | 5, 6 | 853 | |||
XM_017001554.1 | XP_016857043.1 | 4, 5 | 868 | |||
XM_017001555.1 | XP_016857044.1 | 4, 5 | 871 | |||
XM_017001556.1 | XP_016857045.1 | 3, 4 | 1334 | |||
GenBank mRNA | AK000515.1 | - | 3, 4 | 643 | ||
AK299377.1 | - | 3, 4 | 735 | |||
AK316093.1 | - | 3, 4 | 692 | |||
BC063843.1 | - | 3, 4 | 744 | 6501461 |