Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BANP SIRNA Assays ›
Gene Symbol:
BANP
Gene Name:
BTG3 associated nuclear protein
Gene Aliases:
BEND1, SMAR1, SMARBP1
Chromosome Location:
Chr.16: 87949216 - 88077318 on Build GRCh38
UniGene ID:
Hs.461705
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s29888
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BANP
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001173539.1
NP_001167010.1
12
1545
NM_001173540.1
NP_001167011.1
12
1521
NM_001173541.1
NP_001167012.1
11
1428
NM_001173542.1
NP_001167013.1
12
1593
NM_001173543.1
NP_001167014.1
13
1656
NM_017869.3
NP_060339.2
11
1437
NM_079837.2
NP_524576.2
12
1503
XM_005256008.3
XP_005256065.1
12
1570
XM_005256009.3
XP_005256066.1
12
1552
XM_011523170.2
XP_011521472.1
13
1636
XM_011523172.2
XP_011521474.1
14
1881
XM_011523174.2
XP_011521476.1
13
2407
XM_011523175.2
XP_011521477.1
13
2038
XM_011523176.2
XP_011521478.1
13
2100
XM_011523177.2
XP_011521479.1
13
2608
XM_011523178.2
XP_011521480.1
13
1627
XM_011523179.2
XP_011521481.1
13
1618
XM_011523180.2
XP_011521482.1
14
1700
XM_011523181.2
XP_011521483.1
12
1561
XM_011523183.2
XP_011521485.1
13
1613
XM_011523184.2
XP_011521486.1
14
1651
XM_011523186.2
XP_011521488.1
9
1105
XM_017023378.1
XP_016878867.1
14
1740
XM_017023379.1
XP_016878868.1
13
2011
XM_017023380.1
XP_016878869.1
12
1494
XM_017023381.1
XP_016878870.1
11
1446
XM_017023382.1
XP_016878871.1
9
1079
XM_017023383.1
XP_016878872.1
10
1217
GenBank mRNA
AK001039.1
-
11
1425
AK094158.1
-
13
1657
AK125232.1
-
12
1501
AK293472.1
-
12
1507
AK297945.1
-
12
1475
AK315089.1
-
11
1426
AK315535.1
-
12
1593
AL157429.1
-
2
948
BC009424.1
-
11
1382
3530821
KU178664.1
-
10
1285
KU178665.1
-
12
1484
KU178666.1
-
12
1464
KU178667.1
-
11
1402
KU178668.1
-
11
1393
KU178669.1
-
11
1342
KU178670.1
-
10
1298
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
cell cycle
multicellular organism development
covalent chromatin modification
protein localization to nucleus
negative regulation of protein catabolic process
positive regulation of transcription, DNA-templated
regulation of signal transduction by p53 class mediator
p53 binding
DNA binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-7745c474b-6cdhd:80/100.66.78.96:80.
git-commit: c3c9db9cc82507548abdd0ae6a4ade3f81f66292
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.20.0-2024.11.45-1.0