Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHGB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001819.2 | NP_001810.2 | 4 | 1974 | ||
GenBank mRNA | AB209712.1 | - | 4 | 1771 | ||
AK289386.1 | - | 4 | 1794 | |||
AK293536.1 | - | 4 | 1528 | |||
AK312237.1 | - | 4 | 1794 | |||
BC000375.2 | - | 4 | 1772 | 2820258 | ||
CR456726.1 | - | 4 | 1668 | |||
Y00064.1 | - | 4 | 1780 |