Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM118A SIRNA Assays ›
Gene Symbol:
FAM118A
Gene Name:
family with sequence similarity 118 member A
Gene Aliases:
C22orf8
Chromosome Location:
Chr.22: 45309200 - 45341955 on Build GRCh38
UniGene ID:
Hs.265018
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s29975
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM118A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001104595.1
NP_001098065.1
4
896
NM_017911.2
NP_060381.2
3
569
XM_011530247.2
XP_011528549.1
3
673
XM_011530254.2
XP_011528556.1
4
592
XM_011530256.1
XP_011528558.1
3
461
XM_017028841.1
XP_016884330.1
3
458
XM_017028842.1
XP_016884331.1
3
461
XM_017028843.1
XP_016884332.1
3
458
XM_017028844.1
XP_016884333.1
3
461
XM_017028845.1
XP_016884334.1
4
658
XM_017028846.1
XP_016884335.1
3
542
XM_017028847.1
XP_016884336.1
3
1186
XM_017028848.1
XP_016884337.1
3
584
XM_017028849.1
XP_016884338.1
3
420
XM_017028850.1
XP_016884339.1
3
543
XM_017028851.1
XP_016884340.1
3
1174
XM_017028852.1
XP_016884341.1
3
580
XM_017028853.1
XP_016884342.1
3
461
XM_017028854.1
XP_016884343.1
3
543
XM_017028855.1
XP_016884344.1
4
593
XM_017028856.1
XP_016884345.1
4
593
XM_017028857.1
XP_016884346.1
3
542
XM_017028858.1
XP_016884347.1
3
543
XM_017028859.1
XP_016884348.1
3
542
XM_017028860.1
XP_016884349.1
3
543
XM_017028861.1
XP_016884350.1
3
543
GenBank mRNA
AK000642.1
-
3
539
AK125027.1
-
4
1114
BC013696.1
-
3
326
3860694
CR456403.1
-
2
295
DB153788.1
-
3
355
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-4vtwv:80/10.239.167.181:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline