Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HERC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165136.1 | NP_001158608.1 | 13 | 1788 | ||
NM_017912.3 | NP_060382.3 | 13 | 1788 | |||
XM_005263083.3 | XP_005263140.1 | 13 | 1985 | |||
XM_011532053.2 | XP_011530355.1 | 13 | 1987 | |||
XM_017008333.1 | XP_016863822.1 | 12 | 1346 | |||
GenBank mRNA | AF336798.1 | - | 13 | 1788 | ||
AK097168.1 | - | 7 | 603 | |||
AK295832.1 | - | 13 | 1698 | |||
AL832670.1 | - | 1 | 337 | |||
AL833664.1 | - | 2 | 116 | |||
AY653201.1 | - | 13 | 1788 | |||
BC042047.1 | - | 13 | 1664 | 5531519 | ||
BX647121.1 | - | 13 | 1464 |