Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HERC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165136.1 | NP_001158608.1 | 12 | 1657 | ||
NM_017912.3 | NP_060382.3 | 12 | 1657 | |||
XM_005263083.3 | XP_005263140.1 | 12 | 1854 | |||
XM_011532053.2 | XP_011530355.1 | 12 | 1856 | |||
XM_017008333.1 | XP_016863822.1 | 11 | 1215 | |||
GenBank mRNA | AF336798.1 | - | 12 | 1657 | ||
AK097168.1 | - | 6 | 472 | |||
AK295832.1 | - | 12 | 1567 | |||
AY653201.1 | - | 12 | 1657 | |||
AY653202.1 | - | 11 | 1535 | |||
AY653203.1 | - | 10 | 1398 | |||
BC042047.1 | - | 12 | 1533 | 5531519 | ||
BX647121.1 | - | 12 | 1333 |