Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: STX17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017919.2 | NP_060389.2 | 5 | 597 | ||
XM_011518820.2 | XP_011517122.1 | 5 | 680 | |||
XM_011518821.2 | XP_011517123.1 | 5 | 928 | |||
XM_011518823.2 | XP_011517125.1 | 3 | 418 | |||
XM_017014875.1 | XP_016870364.1 | 6 | 774 | |||
GenBank mRNA | AK000658.1 | - | 5 | 546 | ||
AK095262.1 | - | 5 | 1208 | |||
AL833620.1 | - | 5 | 1109 | |||
AL834371.1 | - | 1 | 34 | |||
BC070269.1 | - | 5 | 540 | |||
BC101564.1 | - | 4 | 534 | 8069070 | ||
BC101566.1 | - | 4 | 534 | 8069072 | ||
BC111009.1 | - | 5 | 557 | 5768578 |