Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTCD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017952.5 | NP_060422.4 | 15, 16 | 1301 | ||
GenBank mRNA | AK315676.1 | - | 9, 10 | 755 | ||
AL832520.1 | - | 4, 5 | 305 | |||
AY277598.1 | - | 3, 4 | 269 | |||
BC001758.1 | - | 4, 5 | 375 | |||
BC011832.2 | - | 15, 16 | 1236 | 3939273 |