Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PLEKHB2 SIRNA Assays ›
Gene Symbol:
PLEKHB2
Gene Name:
pleckstrin homology domain containing B2
Gene Aliases:
EVT2
Chromosome Location:
Chr.2: 131104847 - 131149852 on Build GRCh38
UniGene ID:
Hs.469944
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s30055
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PLEKHB2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001100623.1
NP_001094093.1
4
799
NM_001267062.1
NP_001253991.1
4
799
NM_001267063.1
NP_001253992.1
4
799
NM_001267064.1
NP_001253993.1
4
799
NM_001267065.1
NP_001253994.1
4
799
NM_001267066.1
NP_001253995.1
4
799
NM_001267067.1
NP_001253996.1
4
799
NM_001267068.1
NP_001253997.1
4
799
NM_001309448.1
NP_001296377.1
3
754
NM_001309451.1
NP_001296380.1
3
754
NM_001309452.1
NP_001296381.1
5
897
NM_017958.2
NP_060428.2
4
799
XM_011511385.2
XP_011509687.1
4
338
XM_011511386.2
XP_011509688.1
4
341
XM_017004408.1
XP_016859897.1
3
267
XM_017004409.1
XP_016859898.1
3
267
XM_017004410.1
XP_016859899.1
3
267
XM_017004411.1
XP_016859900.1
5
1403
XM_017004412.1
XP_016859901.1
5
1403
XM_017004413.1
XP_016859902.1
4
317
GenBank mRNA
AK000790.1
-
4
310
AK093730.1
-
4
1034
AK123710.1
-
4
756
AK126289.1
-
4
799
AK223338.1
-
4
310
AK293880.1
-
4
310
AK296008.1
-
3
263
AK302770.1
-
4
344
BC001428.1
-
4
293
3139876
BC013991.2
-
4
292
3639839
BC050695.1
-
4
294
6162960
BP362174.1
-
3
265
BQ052811.1
-
4
291
BQ649140.1
-
4
308
BU174451.1
-
5
378
DB294071.1
-
4
312
DC303684.1
-
4
317
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell differentiation
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-cs4xr:80/10.239.162.42:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0