Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC52A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104577.1 | NP_001098047.1 | 5 | 2020 | ||
NM_017986.3 | NP_060456.3 | 5 | 1986 | |||
XM_011523951.1 | XP_011522253.1 | 5 | 2008 | |||
GenBank mRNA | AB362533.1 | - | 5 | 1980 | ||
AB362534.1 | - | 4 | 2077 | |||
AK000922.1 | - | 5 | 1579 | |||
AK023570.1 | - | 4 | 2096 | |||
AK123566.1 | - | 4 | 2600 | |||
AY070775.1 | - | 4 | 1307 | |||
BC060810.1 | - | 5 | 1980 | 30348722 | ||
BC092473.1 | - | 5 | 1538 | 30341994 |
Set Membership: |
G-protein coupled receptor |