Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FANCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114636.1 | NP_001108108.1 | 8 | 664 | ||
NM_018062.3 | NP_060532.2 | 8 | 649 | |||
XM_005264395.3 | XP_005264452.1 | 8 | 649 | |||
XM_005264397.4 | XP_005264454.1 | 4 | 368 | |||
XM_011532939.2 | XP_011531241.1 | 9 | 709 | |||
XM_011532940.2 | XP_011531242.1 | 9 | 694 | |||
XM_011532941.1 | XP_011531243.1 | 9 | 709 | |||
XM_011532942.2 | XP_011531244.1 | 8 | 664 | |||
XM_011532944.2 | XP_011531246.1 | 5 | 386 | |||
XM_011532945.2 | XP_011531247.1 | 4 | 354 | |||
XM_017004414.1 | XP_016859903.1 | 9 | 694 | |||
XM_017004416.1 | XP_016859905.1 | 4 | 368 | |||
GenBank mRNA | AK001197.1 | - | 8 | 615 | ||
AK297736.1 | - | 6 | 458 | |||
AK309671.1 | - | 5 | 464 | |||
BC009042.1 | - | 8 | 619 | 3870828 | ||
BC054517.1 | - | 8 | 624 | 5180184 |