Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ANO10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204831.1 | NP_001191760.1 | 6 | 1298 | ||
NM_001204832.1 | NP_001191761.1 | 5 | 1100 | |||
NM_001204833.1 | NP_001191762.1 | 4 | 965 | |||
NM_018075.3 | NP_060545.3 | 6 | 1298 | |||
XM_011533885.2 | XP_011532187.2 | 6 | 1264 | |||
XM_011533887.2 | XP_011532189.1 | 6 | 1264 | |||
XM_011533889.2 | XP_011532191.1 | 6 | 1264 | |||
XM_011533890.2 | XP_011532192.1 | 6 | 1264 | |||
XM_017006716.1 | XP_016862205.1 | 6 | 1264 | |||
XM_017006717.1 | XP_016862206.1 | 6 | 1216 | |||
XM_017006718.1 | XP_016862207.1 | 6 | 1243 | |||
XM_017006719.1 | XP_016862208.1 | 5 | 1018 | |||
XM_017006720.1 | XP_016862209.1 | 6 | 1243 | |||
XM_017006721.1 | XP_016862210.1 | 5 | 1027 | |||
XM_017006722.1 | XP_016862211.1 | 5 | 1000 | |||
GenBank mRNA | AK096302.1 | - | 6 | 1298 | ||
AK131223.1 | - | 5 | 1016 | |||
AK292368.1 | - | 6 | 1183 | |||
AK295969.1 | - | 4 | 900 | |||
AK297949.1 | - | 6 | 1241 | |||
AL832508.1 | - | 6 | 1165 |