Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANO10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204831.1 | NP_001191760.1 | 11 | 1861 | ||
NM_001204832.1 | NP_001191761.1 | 10 | 1663 | |||
NM_001204833.1 | NP_001191762.1 | 9 | 1528 | |||
NM_001204834.1 | NP_001191763.1 | 10 | 1291 | |||
NM_018075.3 | NP_060545.3 | 11 | 1861 | |||
XM_011533885.2 | XP_011532187.2 | 12 | 1944 | |||
XM_011533887.2 | XP_011532189.1 | 12 | 1944 | |||
XM_011533889.2 | XP_011532191.1 | 12 | 1944 | |||
XM_011533890.2 | XP_011532192.1 | 12 | 1944 | |||
XM_017006716.1 | XP_016862205.1 | 12 | 1944 | |||
XM_017006717.1 | XP_016862206.1 | 12 | 1896 | |||
XM_017006718.1 | XP_016862207.1 | 11 | 1806 | |||
XM_017006719.1 | XP_016862208.1 | 11 | 1698 | |||
XM_017006720.1 | XP_016862209.1 | 11 | 1806 | |||
XM_017006721.1 | XP_016862210.1 | 10 | 1590 | |||
XM_017006722.1 | XP_016862211.1 | 10 | 1563 | |||
GenBank mRNA | AK096302.1 | - | 11 | 1861 | ||
AK131223.1 | - | 10 | 1579 | |||
AK292368.1 | - | 11 | 1746 | |||
AK295969.1 | - | 9 | 1463 | |||
AK297949.1 | - | 11 | 1804 | |||
AL832508.1 | - | 11 | 1728 |