Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290020.1 | NP_001276949.1 | 9 | 1354 | ||
NM_001312689.1 | NP_001299618.1 | 4 | 571 | |||
NM_018076.4 | NP_060546.2 | 9 | 1307 | |||
XM_011519527.1 | XP_011517829.1 | 9 | 1354 | |||
XM_011519528.1 | XP_011517830.1 | 9 | 2004 | |||
XM_011519533.2 | XP_011517835.1 | 4 | 528 | |||
XM_011519534.2 | XP_011517836.1 | 4 | 661 | |||
XM_011519535.2 | XP_011517837.1 | 3 | 413 | |||
XM_017016368.1 | XP_016871857.1 | 9 | 1354 | |||
XM_017016369.1 | XP_016871858.1 | 9 | 1354 | |||
XM_017016370.1 | XP_016871859.1 | 9 | 1354 | |||
XM_017016371.1 | XP_016871860.1 | 9 | 1354 | |||
GenBank mRNA | AK001238.1 | - | 7 | 894 | ||
AK001679.1 | - | 9 | 1307 | |||
AK292521.1 | - | 9 | 1310 | |||
AK301708.1 | - | 4 | 482 | |||
AK302419.1 | - | 4 | 903 | |||
AK302627.1 | - | 4 | 571 | |||
BC140846.1 | - | 8 | 1235 | 9021714 |
Gene Symbol: ARMC4P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125237.1 | - | 469 |