Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018105.2 | NP_060575.1 | 1 | 287 | ||
NM_199003.1 | NP_945354.1 | 1 | 287 | |||
GenBank mRNA | AK001339.1 | - | 1 | 281 | ||
AK223231.1 | - | 1 | 266 | |||
AL832077.1 | - | 1 | 261 | |||
BC021721.2 | - | 1 | 261 | 4795229 | ||
BG497522.1 | - | 1 | 288 | |||
CD639319.1 | - | 1 | 269 | |||
CD709111.1 | - | 1 | 297 | |||
CR457256.1 | - | 1 | 50 |