Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NADSYN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018161.4 | NP_060631.2 | 8 | 769 | ||
GenBank mRNA | AB091316.1 | - | 8 | 643 | ||
AK001493.1 | - | 8 | 710 | |||
AK022436.1 | - | 8 | 739 | |||
AK225700.1 | - | 8 | 723 | |||
BC003638.2 | - | 8 | 732 | 2966503 | ||
BC003666.2 | - | 8 | 732 | 2966503 | ||
CR457286.1 | - | 8 | 643 |