Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PBRM1 SIRNA Assays ›
Gene Symbol:
PBRM1
Gene Name:
polybromo 1
Gene Aliases:
BAF180, PB1
Chromosome Location:
Chr.3: 52545352 - 52685933 on Build GRCh38
UniGene ID:
Hs.189920
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s30402
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PBRM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_018313.4
NP_060783.3
14
1588
XM_005265279.4
XP_005265336.1
15
1816
XM_005265280.3
XP_005265337.1
14
1633
XM_005265282.3
XP_005265339.1
15
1775
XM_005265283.3
XP_005265340.1
14
1630
XM_011533900.2
XP_011532202.1
17
1982
XM_011533902.2
XP_011532204.1
16
1914
XM_011533903.2
XP_011532205.1
16
1919
XM_017006725.1
XP_016862214.1
16
1889
XM_017006726.1
XP_016862215.1
17
1999
XM_017006727.1
XP_016862216.1
17
1944
XM_017006728.1
XP_016862217.1
16
1940
XM_017006729.1
XP_016862218.1
16
2348
XM_017006730.1
XP_016862219.1
17
1935
XM_017006731.1
XP_016862220.1
16
1918
XM_017006732.1
XP_016862221.1
16
1914
XM_017006733.1
XP_016862222.1
16
2308
XM_017006734.1
XP_016862223.1
16
1881
XM_017006735.1
XP_016862224.1
16
2347
XM_017006736.1
XP_016862225.1
16
2348
XM_017006737.1
XP_016862226.1
15
2222
XM_017006738.1
XP_016862227.1
15
1843
XM_017006739.1
XP_016862228.1
15
2219
XM_017006740.1
XP_016862229.1
15
1760
XM_017006741.1
XP_016862230.1
16
1896
XM_017006742.1
XP_016862231.1
16
1919
XM_017006743.1
XP_016862232.1
16
2348
XM_017006744.1
XP_016862233.1
16
1885
XM_017006745.1
XP_016862234.1
15
2194
XM_017006746.1
XP_016862235.1
15
2191
XM_017006747.1
XP_016862236.1
15
2253
XM_017006748.1
XP_016862237.1
15
2194
XM_017006749.1
XP_016862238.1
15
1760
XM_017006750.1
XP_016862239.1
14
1648
XM_017006751.1
XP_016862240.1
16
1896
XM_017006752.1
XP_016862241.1
16
2348
XM_017006753.1
XP_016862242.1
16
2347
XM_017006754.1
XP_016862243.1
16
1914
XM_017006755.1
XP_016862244.1
16
1896
XM_017006756.1
XP_016862245.1
16
1896
XM_017006757.1
XP_016862246.1
14
1619
XM_017006758.1
XP_016862247.1
14
1750
XM_017006759.1
XP_016862248.1
16
1896
XM_017006760.1
XP_016862249.1
16
1914
XM_017006761.1
XP_016862250.1
16
1923
XM_017006762.1
XP_016862251.1
16
1914
XM_017006763.1
XP_016862252.1
17
1973
XM_017006764.1
XP_016862253.1
16
1896
XM_017006765.1
XP_016862254.1
15
1775
XM_017006766.1
XP_016862255.1
16
1903
GenBank mRNA
AF177387.1
-
13
1477
AF197569.1
-
14
1570
AF225870.1
-
12
1381
AF225871.1
-
13
1477
AF225872.1
-
13
1477
AK056541.1
-
12
1306
AY281068.1
-
13
1476
BC115009.1
-
13
1492
7962003
BC115010.1
-
13
1493
BC115011.1
-
13
1493
7962010
BC129934.1
-
13
1477
40082629
BC129935.1
-
13
1477
40082630
BU171600.1
-
6
683
Back To Top
More Information
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
chromatin remodeling
transcription, DNA-templated
regulation of transcription, DNA-templated
mitotic nuclear division
negative regulation of cell proliferation
covalent chromatin modification
DNA binding
chromatin binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-xbcvc:80/10.239.164.135:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline