Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UQCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184977.1 | NP_001171906.1 | 7 | 678 | ||
NM_018244.4 | NP_060714.3 | 9 | 882 | |||
NM_199487.2 | NP_955781.2 | 8 | 804 | |||
XM_011528877.1 | XP_011527179.1 | 10 | 829 | |||
XM_011528878.1 | XP_011527180.1 | 8 | 691 | |||
XM_011528879.1 | XP_011527181.1 | 9 | 733 | |||
XM_011528880.1 | XP_011527182.1 | 9 | 791 | |||
XM_011528881.2 | XP_011527183.1 | 8 | 813 | |||
XM_011528882.1 | XP_011527184.1 | 8 | 667 | |||
XM_011528883.1 | XP_011527185.1 | 7 | 593 | |||
XM_011528884.1 | XP_011527186.1 | 7 | 544 | |||
GenBank mRNA | AK001712.1 | - | 9 | 713 | ||
AK022650.1 | - | 8 | 635 | |||
AK022742.1 | - | 9 | 763 | |||
AK225281.1 | - | 9 | 722 | |||
AK225557.1 | - | 9 | 713 | |||
AK293149.1 | - | 4 | 287 | |||
AK293516.1 | - | 7 | 509 | |||
AK295379.1 | - | 7 | 558 | |||
AK300130.1 | - | 7 | 582 | |||
AK302133.1 | - | 5 | 302 | |||
AY050659.1 | - | 9 | 882 | |||
AY050660.1 | - | 7 | 604 | |||
BC105791.1 | - | 9 | 717 | 6458054 | ||
BC119013.1 | - | 9 | 748 | 40085306 | ||
BC122862.1 | - | 9 | 748 | 40085305 | ||
BM919736.1 | - | 9 | 735 | |||
BX537568.1 | - | 1 | 331 |