Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UQCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018244.4 | NP_060714.3 | 3 | 368 | ||
NM_199487.2 | NP_955781.2 | 3 | 368 | |||
XM_011528877.1 | XP_011527179.1 | 4 | 315 | |||
XM_011528879.1 | XP_011527181.1 | 3 | 219 | |||
XM_011528880.1 | XP_011527182.1 | 3 | 277 | |||
XM_011528882.1 | XP_011527184.1 | 3 | 226 | |||
GenBank mRNA | AK001712.1 | - | 3 | 199 | ||
AK022650.1 | - | 3 | 199 | |||
AK022742.1 | - | 4 | 322 | |||
AK225281.1 | - | 3 | 208 | |||
AK225557.1 | - | 3 | 199 | |||
AK300130.1 | - | 3 | 199 | |||
AK302133.1 | - | 3 | 199 | |||
AU280736.1 | - | 3 | 199 | |||
AY050659.1 | - | 3 | 368 | |||
AY050660.1 | - | 2 | 171 | |||
BC105791.1 | - | 3 | 203 | 6458054 | ||
BC119013.1 | - | 3 | 234 | 40085306 | ||
BC122862.1 | - | 3 | 234 | 40085305 | ||
BM919736.1 | - | 3 | 221 |