Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306077.1 | NP_001293006.1 | 2, 3 | 3067 | ||
NM_018269.3 | NP_060739.2 | 2, 3 | 339 | |||
GenBank mRNA | AB073609.1 | - | 2, 3 | 290 | ||
AB158319.1 | - | 2, 3 | 280 | |||
AF087863.1 | - | 2, 3 | 286 | |||
AF403478.1 | - | 1, 2 | 100 | |||
AK001775.1 | - | 2, 3 | 291 | |||
AK127473.1 | - | 2, 3 | 3062 | |||
AK222467.1 | - | 2, 3 | 285 | |||
AK225278.1 | - | 2, 3 | 287 | |||
BC001467.2 | - | 2, 3 | 280 | 3138914 | ||
CB129674.1 | - | 2, 3 | 339 | |||
DA559726.1 | - | 2, 3 | 317 |