Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THNSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244676.1 | NP_001231605.1 | 4 | 653 | ||
NM_001244678.1 | NP_001231607.1 | 2 | 223 | |||
NM_018271.4 | NP_060741.3 | 3 | 491 | |||
XM_005264400.4 | XP_005264457.1 | 4 | 603 | |||
XM_005264401.4 | XP_005264458.1 | 4 | 572 | |||
XM_005264402.3 | XP_005264459.1 | 3 | 2392 | |||
XM_005264403.4 | XP_005264460.1 | 3 | 2392 | |||
XM_005264405.4 | XP_005264462.1 | 3 | 2392 | |||
XM_006712041.3 | XP_006712104.1 | 3 | 2392 | |||
XM_006712042.3 | XP_006712105.1 | 3 | 2392 | |||
XM_006712043.2 | XP_006712106.1 | 3 | 2392 | |||
XM_006712044.3 | XP_006712107.1 | 3 | 2392 | |||
XM_011532953.2 | XP_011531255.1 | 3 | 2392 | |||
XM_011532954.2 | XP_011531256.1 | 2 | 2197 | |||
XM_011532955.2 | XP_011531257.1 | 3 | 337 | |||
GenBank mRNA | AK001778.1 | - | 3 | 294 | ||
AK095303.1 | - | 3 | 2171 | |||
AK289486.1 | - | 3 | 323 | |||
AK315831.1 | - | 2 | 149 | |||
AL832746.1 | - | 1 | 23 | |||
BC035315.1 | - | 3 | 322 | |||
BC047758.2 | - | 1 | 62 | |||
BC064423.1 | - | 4 | 550 | 5752279 | ||
BI765911.1 | - | 3 | 405 |